Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-(trifluoromethyl)benzenesulfonamide, 97%

$53.49 - $150.68

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzenesulfonamide
CAS 146533-47-3
Molecular Formula C7H5ClF3NO2S
Formula Weight 259.63
MDL Number MFCD00179384
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3263703

  SDS
alfa aesar™
H3263703
1g Each for $150.68

AAH32637MD

  SDS
alfa aesar™
H32637MD
250mg Each for $53.49
Description & Specifications

Specifications

Formula Weight 259.63
Quantity 1g
CAS 146533-47-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 168° to 171°C
Assay 97%
Molecular Formula C7H5ClF3NO2S
MDL Number MFCD00179384
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzenesulfonamide
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.