missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-(trifluoromethyl)aniline, 98%

Manufacturer:  Alfa Aesar™ A1224214

 View more versions of this product

Catalog No. AAA1224214



Specifications

Specifications

2-Chloro-4- (trifluoromethyl)aniline
214°C to 218°C
39885-50-2
MFCD00042563
4179703
MBBUTABXEITVNY-UHFFFAOYSA-N
2-chloro-4-(trifluoromethyl)aniline
162001
98%
103°C (217°F)
25g
C7H5ClF3N
UN2810
2-chloro-4-trifluoromethyl aniline, 4-amino-3-chlorobenzotrifluoride, 4-amino-5-chloro-benzotrifluoride, benzenamine, 2-chloro-4-trifluoromethyl, 3-chloro-4-aminobenzotrifluoride, 2-chloro-4-trifluoromethyl-phenylamine, 2-chloro-4-trifluoromethyl benzenamine, 2-chloro-4-trifluoromethyl phenylamine, 4-amino-3-chloro-benzotrifluoride, pubchem2474
C1=CC(=C(C=C1C(F)(F)F)Cl)N
195.569
195.57

Chemical Identifiers

39885-50-2
195.569
MBBUTABXEITVNY-UHFFFAOYSA-N
162001
C1=CC(=C(C=C1C(F)(F)F)Cl)N
C7H5ClF3N
MFCD00042563
2-chloro-4-trifluoromethyl aniline, 4-amino-3-chlorobenzotrifluoride, 4-amino-5-chloro-benzotrifluoride, benzenamine, 2-chloro-4-trifluoromethyl, 3-chloro-4-aminobenzotrifluoride, 2-chloro-4-trifluoromethyl-phenylamine, 2-chloro-4-trifluoromethyl benzenamine, 2-chloro-4-trifluoromethyl phenylamine, 4-amino-3-chloro-benzotrifluoride, pubchem2474
2-chloro-4-(trifluoromethyl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

SDS Documents