Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Chloro-4'-(trifluoromethyl)acetanilide, 96%

$59.12 - $224.73

Chemical Identifiers

Chemical Name or Material 2'-Chloro-4'-(trifluoromethyl)acetanilide
CAS 247170-19-0
Molecular Formula C9H7ClF3NO
Formula Weight 237.61
MDL Number MFCD01313231
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2231003

 
alfa aesar™
B2231003
1g Each for $59.12

AAB2231006

 
alfa aesar™
B2231006
5g Each for $224.73
Description & Specifications

Specifications

Formula Weight 237.61
Chemical Name or Material 2'-Chloro-4'-(trifluoromethyl)acetanilide
Assay 96%
Molecular Formula C9H7ClF3NO
MDL Number MFCD01313231
CAS 247170-19-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 134° to 138°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.