Please login to your online account to display your discounted pricing

2-Chloro-4-(trifluoromethoxy)aniline, 95%, ACROS Organics™

$129.08 - $129.08

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-(trifluoromethoxy)aniline
CAS 69695-61-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H5ClF3NO
Formula Weight 211.57
MDL Number MFCD01631704
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC350210050

  SDS
thermo fisher scientific
AC350210050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350210010

  SDS
thermo fisher scientific
350210010
Glass bottle 1g Each for $129.08
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 211.57
Quantity 5g
Color Pale yellow to yellow-brown
Density 98g/mL
CAS Min % 94.0
CAS Max % 100.0
Refractive Index 1.4790 to 1.4830 (20°C, 589nm)
Chemical Name or Material 2-Chloro-4-(trifluoromethoxy)aniline
Molecular Formula C7H5ClF3NO
CAS 69695-61-0
Assay Percent Range 95%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD01631704
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.