Please login to your online account to display your discounted pricing

2-Chloro-4-(trifluoromethoxy)aniline, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  350210010

 View more versions of this product

Catalog No. AC350210010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 211.57
Quantity 1g
Color Pale yellow to yellow-brown
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass bottle
CAS 69695-61-0
Assay Percent Range 95%
Chemical Name or Material 2-Chloro-4-(trifluoromethoxy)aniline
Molecular Formula C7H5ClF3NO
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD01631704
Refractive Index 1.4790 to 1.4830 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2-Chloro-4-(trifluoromethoxy)aniline
CAS 69695-61-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H5ClF3NO
Formula Weight 211.57
MDL Number MFCD01631704
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents