missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4-picoline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC368840050

 View more versions of this product

Catalog No. AC368840050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Chloro-4-picoline
89°C
Glass bottle
1.151
Colorless to Yellow
3678-62-4
100.0
C6H6ClN
20,241
MZVSTDHRRYQFGI-UHFFFAOYSA-N
2-chloro-4-methylpyridine
77248
Liquid
1.1510g/mL
Authentic
1.5280 to 1.5300 (20°C, 589nm)
194°C to 195°C
5mL
97.5
97.5% min. (GC)
MFCD00023418
2-chloro-4-methyl pyridine, 2-chloro-4-methyl-pyridine, 2-chloro-4-picoline, 2-chloro-gamma-picoline, 4-picoline, 2-chloro, acmc-209iop, pubchem1192, pyridine, 2-chloro-4-methyl, zlchem 438
CC1=CC(=NC=C1)Cl
127.57
127.57
98%

Chemical Identifiers

3678-62-4
127.57
MZVSTDHRRYQFGI-UHFFFAOYSA-N
77248
CC1=CC(=NC=C1)Cl
C6H6ClN
MFCD00023418
2-chloro-4-methyl pyridine, 2-chloro-4-methyl-pyridine, 2-chloro-4-picoline, 2-chloro-gamma-picoline, 4-picoline, 2-chloro, acmc-209iop, pubchem1192, pyridine, 2-chloro-4-methyl, zlchem 438
2-chloro-4-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 222-951-2

SDS Documents
Product Certifications