missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-4'-phenylacetophenone 99.0+%, TCI America™
$58.06 - $291.98
Chemical Identifiers
CAS | 635-84-7 |
---|---|
Molecular Formula | C14H11ClO |
Molecular Weight (g/mol) | 230.69 |
MDL Number | MFCD00058937 |
InChI Key | IQEIGQFNDLINOT-UHFFFAOYSA-N |
Synonym | 4-Chloroacetylbiphenyl, 4-Phenylphenacyl Chloride |
PubChem CID | 69463 |
IUPAC Name | 1-{[1,1'-biphenyl]-4-yl}-2-chloroethan-1-one |
SMILES | ClCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
Chemical Identifiers
635-84-7 | |
230.69 | |
IQEIGQFNDLINOT-UHFFFAOYSA-N | |
69463 | |
ClCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
C14H11ClO | |
MFCD00058937 | |
4-Chloroacetylbiphenyl, 4-Phenylphenacyl Chloride | |
1-{[1,1'-biphenyl]-4-yl}-2-chloroethan-1-one |
Specifications
635-84-7 | |
White-Yellow | |
MFCD00058937 | |
4-Chloroacetylbiphenyl, 4-Phenylphenacyl Chloride | |
ClCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 | |
230.69 | |
230.69 | |
Crystalline Powder |
126°C | |
C14H11ClO | |
5 g | |
IQEIGQFNDLINOT-UHFFFAOYSA-N | |
1-{[1,1'-biphenyl]-4-yl}-2-chloroethan-1-one | |
69463 | |
≥99.0% (GC,T) | |
2-Chloro-4′-phenylacetophenone |
Safety and Handling
TSCA : Yes