Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-nitropyridine N-oxide, 97%

$141.95 - $559.01

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-nitropyridine N-oxide
CAS 14432-16-7
Molecular Formula C5H3ClN2O3
Formula Weight 174.54
MDL Number MFCD00955617
Beilstein 145697
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2170718

  SDS
alfa aesar™
B2170718
50g Each for $559.01

AAB2170709

  SDS
alfa aesar™
B2170709
10g Each for $141.95
Description & Specifications

Specifications

Formula Weight 174.54
Chemical Name or Material 2-Chloro-4-nitropyridine N-oxide
Odor Odorless
Assay 97%
Molecular Formula C5H3ClN2O3
MDL Number MFCD00955617
CAS 14432-16-7
Beilstein 145697
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 152° to 155°C
Quantity 50g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.