missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-4-nitropyridine, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC443530050
Specifications
2-Chloro-4-nitropyridine | |
5g | |
97.5% min. (GC) | |
2-chlor-4-nitropyridin, 2-chloro-4-nitro pyridine, 2-chloro-4-nitro-pyridine, 2-chloro-4-nitropyridine, 2-chloro-4nitropyridine, 2chloro-4-nitropyridine, acmc-1ccv7, ksc201q1f, pubchem9220, pyridine, 2-chloro-4-nitro | |
C1=CN=C(C=C1[N+](=O)[O-])Cl | |
158.54 | |
158.54 |
52.0°C to 56.0°C | |
23056-36-2 | |
C5H3ClN2O2 | |
LIEPVGBDUYKPLC-UHFFFAOYSA-N | |
2-chloro-4-nitropyridine | |
735152 | |
98% |
Chemical Identifiers
23056-36-2 | |
158.54 | |
2-chlor-4-nitropyridin, 2-chloro-4-nitro pyridine, 2-chloro-4-nitro-pyridine, 2-chloro-4-nitropyridine, 2-chloro-4nitropyridine, 2chloro-4-nitropyridine, acmc-1ccv7, ksc201q1f, pubchem9220, pyridine, 2-chloro-4-nitro | |
2-chloro-4-nitropyridine |
C5H3ClN2O2 | |
LIEPVGBDUYKPLC-UHFFFAOYSA-N | |
735152 | |
C1=CN=C(C=C1[N+](=O)[O-])Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning