Please login to your online account to display your discounted pricing

2-Chloro-4-nitropyridine, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-nitropyridine
Assay Percent Range 98%
Molecular Formula C5H3ClN2O2
Formula Weight 158.54
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC443530050

  SDS
thermo fisher scientific
AC443530050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC443530010

  SDS
thermo fisher scientific
AC443530010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.54
Quantity 5g
Molecular Formula C5H3ClN2O2
CAS 23056-36-2
Chemical Name or Material 2-Chloro-4-nitropyridine
Assay Percent Range 98%