missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-nitropyridine, 98%

$81.73 - $262.16

Chemical Identifiers

CAS 23056-36-2
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.541
MDL Number MFCD00661454
InChI Key LIEPVGBDUYKPLC-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-4-nitro, 2-chloro-4-nitro-pyridine, 2-chlor-4-nitropyridin, pubchem9220, 2-chloro-4nitropyridine, 2chloro-4-nitropyridine, acmc-1ccv7, 2-chloro-4-nitro pyridine, ksc201q1f, 2-chloro-4-nitropyridine
PubChem CID 735152
IUPAC Name 2-chloro-4-nitropyridine
SMILES C1=CN=C(C=C1[N+](=O)[O-])Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1769903 View Documents Alfa Aesar™
L1769903
1g Each for $81.73
 
AAL1769906 View Documents Alfa Aesar™
L1769906
5g Each for $262.16
 
Specifications

Chemical Identifiers

23056-36-2
158.541
LIEPVGBDUYKPLC-UHFFFAOYSA-N
735152
C1=CN=C(C=C1[N+](=O)[O-])Cl
C5H3ClN2O2
MFCD00661454
pyridine, 2-chloro-4-nitro, 2-chloro-4-nitro-pyridine, 2-chlor-4-nitropyridin, pubchem9220, 2-chloro-4nitropyridine, 2chloro-4-nitropyridine, acmc-1ccv7, 2-chloro-4-nitro pyridine, ksc201q1f, 2-chloro-4-nitropyridine
2-chloro-4-nitropyridine

Specifications

50°C to 54°C
23056-36-2
MFCD00661454
pyridine, 2-chloro-4-nitro, 2-chloro-4-nitro-pyridine, 2-chlor-4-nitropyridin, pubchem9220, 2-chloro-4nitropyridine, 2chloro-4-nitropyridine, acmc-1ccv7, 2-chloro-4-nitro pyridine, ksc201q1f, 2-chloro-4-nitropyridine
C1=CN=C(C=C1[N+](=O)[O-])Cl
158.541
158.54
2-Chloro-4-nitropyridine
1g
C5H3ClN2O2
120418
LIEPVGBDUYKPLC-UHFFFAOYSA-N
2-chloro-4-nitropyridine
735152
98%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning