Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-nitrophenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-nitrophenyl isocyanate
CAS 40397-95-3
Molecular Formula C7H3ClN2O3
Formula Weight 198.57
MDL Number MFCD00037063
UN Number UN2811
Beilstein 2841619
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1268503

  SDS
alfa aesar™
L1268503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1268506

  SDS
alfa aesar™
L1268506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.57
Chemical Name or Material 2-Chloro-4-nitrophenyl isocyanate
Health Hazard 2 GHS H Statement
H331-H334-H302-H312-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
MDL Number MFCD00037063
CAS 40397-95-3
Assay 97%
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H3ClN2O3
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
Recommended Storage 2° to 8°C
Melting Point 59° to 63°C
Quantity 1g
Beilstein 2841619
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H312-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

2° to 8°C