missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-nitrobenzonitrile, 98%

$169.32 - $169.32

Chemical Identifiers

CAS 28163-00-0
Molecular Formula C7H3ClN2O2
Formula Weight 182.57
MDL Number MFCD00052203
Synonym 3-Chloro-4-cyanonitrobenzene
Chemical Name or Material 2-Chloro-4-nitrobenzonitrile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1197706 SDS Alfa Aesar™
A1197706
5g Each for $169.32
AAA1197703 SDS Alfa Aesar™
A1197703
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

28163-00-0
182.57
3-Chloro-4-cyanonitrobenzene
C7H3ClN2O2
MFCD00052203
2-Chloro-4-nitrobenzonitrile

Specifications

78° to 82°C
28163-00-0
182.57
3-Chloro-4-cyanonitrobenzene
98%
5g
C7H3ClN2O2
MFCD00052203
2-Chloro-4-nitrobenzonitrile

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning