missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4-nitrobenzoic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 109655000

 View more versions of this product

Catalog No. AC109655000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Chloro-4-nitrobenzoic acid
99-60-5
100.0
97.5% min. (HPLC)
C7H4ClNO4
MFCD00007209
09, 404
Solubility in water: 1g/L (20 c)
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)O
7448
98%
98%
97.5
Authentic
Plastic bottle
ClC6H3(NO2)CO2H
500g
2-chloro-4-nitrobenzoic acid, benzoic acid, 2-chloro-4-nitro, kyselina 2-chloro-4-nitrobenzoova, kyselina 2-chloro-4-nitrobenzoova czech, 2-chloro-4-nitrobenzoic, 2-chloro-4-nitro-benzoic acid, 2-chloro-4-nitro benzoic acid, 2-chloro-4-nitrobenzoicacid, pubchem20257, acmc-209sde
QAYNSPOKTRVZRC-UHFFFAOYSA-N
201.57
201.57

Chemical Identifiers

99-60-5
201.57
QAYNSPOKTRVZRC-UHFFFAOYSA-N
7448
C7H4ClNO4
MFCD00007209
2-chloro-4-nitrobenzoic acid, benzoic acid, 2-chloro-4-nitro, kyselina 2-chloro-4-nitrobenzoova, kyselina 2-chloro-4-nitrobenzoova czech, 2-chloro-4-nitrobenzoic, 2-chloro-4-nitro-benzoic acid, 2-chloro-4-nitro benzoic acid, 2-chloro-4-nitrobenzoicacid, pubchem20257, acmc-209sde
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-771-0

RTECSNumber : DG5425000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only