Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-nitrobenzoic acid, 98%

$29.39 - $432.54

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-nitrobenzoic acid
CAS 99-60-5
Molecular Formula C7H4ClNO4
Formula Weight 201.57
MDL Number MFCD00007209
Beilstein 1912836
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA116020E

 
alfa aesar™
A116020E
2500g Each for $432.54

AAA1160236

 
alfa aesar™
A1160236
500g Each for $108.07

AAA1160222

 
alfa aesar™
A1160222
100g Each for $29.39
Description & Specifications

Specifications

Formula Weight 201.57
Chemical Name or Material 2-Chloro-4-nitrobenzoic acid
TSCA TSCA
Odor Odorless
Assay 98%
Molecular Formula C7H4ClNO4
MDL Number MFCD00007209
CAS 99-60-5
Beilstein 1912836
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P280a-P305+P351+P338-P337+P313
Wear protective gloves and eye/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
RTECS Number DG5425000
Melting Point 140° to 144°C
Quantity 2500g
Safety and Handling

Danger

GHS P Statement
P280a-P305+P351+P338-P337+P313
Wear protective gloves and eye/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.