missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-4-methylsulphonylbenzoic acid, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 425010010
Specifications
2-Chloro-4-methylsulphonylbenzoic acid | |
53250-83-2 | |
100.0 | |
Pink or White to Yellow | |
Authentic | |
Glass bottle | |
MFCD00216496 | |
2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa | |
CTTWSFIIFMWHLQ-UHFFFAOYSA-M | |
2-chloro-4-methylsulfonylbenzoic acid | |
735863 | |
95% |
95% | |
94.0 | |
192.0°C to 193.0°C | |
5% max. | |
95% | |
C8H6ClO4S | |
1 g | |
Solubility in water: insoluble | |
CS(=O)(=O)C1=CC=C(C([O-])=O)C(Cl)=C1 | |
233.64 | |
234.66 | |
Powder |
Chemical Identifiers
53250-83-2 | |
233.64 | |
CTTWSFIIFMWHLQ-UHFFFAOYSA-M | |
735863 | |
CS(=O)(=O)C1=CC=C(C([O-])=O)C(Cl)=C1 |
C8H6ClO4S | |
MFCD00216496 | |
2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa | |
2-chloro-4-methylsulfonylbenzoic acid |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
TSCA : TSCA
RUO – Research Use Only