missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-4-methylsulphonylaniline, 95%, ACROS Organics™

$167.60 - $167.60

Chemical Identifiers

CAS 13244-35-4
Molecular Formula C7H8ClNO2S
Molecular Weight (g/mol) 205.656
MDL Number MFCD00152349
InChI Key VLMRGLCBIFWPGL-UHFFFAOYSA-N
Synonym 2-chloro-4-methylsulfonyl aniline, 2-chloro-4-mesylaniline, 2-chloro-4-methylsulphonylaniline, 2-chloro-4-methanesulfonylaniline, benzenamine, 2-chloro-4-methylsulfonyl, 4-amino-3-chlorophenylmethylsulphone, 1-amino-2-chloro-4-methylsulfonylbenzene, 4-amino-3-chlorofenylomethylosulfon, 4-amino-3-chlorophenylmethylsulfone, aniline, 2-chloro-4-methylsulfonyl
PubChem CID 25801
IUPAC Name 2-chloro-4-methylsulfonylaniline
SMILES CS(=O)(=O)C1=CC(=C(C=C1)N)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC424990025 SDS View Documents Acros Organics
424990025
2.5g Glass bottle Each for $167.60
 
Specifications

Chemical Identifiers

13244-35-4
205.656
VLMRGLCBIFWPGL-UHFFFAOYSA-N
25801
CS(=O)(=O)C1=CC(=C(C=C1)N)Cl
C7H8ClNO2S
MFCD00152349
2-chloro-4-methylsulfonyl aniline, 2-chloro-4-mesylaniline, 2-chloro-4-methylsulphonylaniline, 2-chloro-4-methanesulfonylaniline, benzenamine, 2-chloro-4-methylsulfonyl, 4-amino-3-chlorophenylmethylsulphone, 1-amino-2-chloro-4-methylsulfonylbenzene, 4-amino-3-chlorofenylomethylosulfon, 4-amino-3-chlorophenylmethylsulfone, aniline, 2-chloro-4-methylsulfonyl
2-chloro-4-methylsulfonylaniline

Specifications

Authentic
Brown-Gray to Purple
2.5g
94.0
95%
MFCD00152349
VLMRGLCBIFWPGL-UHFFFAOYSA-N
2-chloro-4-methylsulfonylaniline
25801
Crystalline Powder
2-chloro-4-methylsulphonylaniline
Glass bottle
191°C to 198°C
13244-35-4
100.0
C7H8ClNO2S
2-chloro-4-methylsulfonyl aniline, 2-chloro-4-mesylaniline, 2-chloro-4-methylsulphonylaniline, 2-chloro-4-methanesulfonylaniline, benzenamine, 2-chloro-4-methylsulfonyl, 4-amino-3-chlorophenylmethylsulphone, 1-amino-2-chloro-4-methylsulfonylbenzene, 4-amino-3-chlorofenylomethylosulfon, 4-amino-3-chlorophenylmethylsulfone, aniline, 2-chloro-4-methylsulfonyl
CS(=O)(=O)C1=CC(=C(C=C1)N)Cl
205.656
205.66
≥94%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.