missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-4-methylsulphonylaniline, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 424990025

Catalog No. AC424990025



Description

Description

Specifications

Specifications

2-chloro-4-methylsulphonylaniline
94.0
Authentic
Glass bottle
MFCD00152349
2-chloro-4-methylsulfonyl aniline, 2-chloro-4-mesylaniline, 2-chloro-4-methylsulphonylaniline, 2-chloro-4-methanesulfonylaniline, benzenamine, 2-chloro-4-methylsulfonyl, 4-amino-3-chlorophenylmethylsulphone, 1-amino-2-chloro-4-methylsulfonylbenzene, 4-amino-3-chlorofenylomethylosulfon, 4-amino-3-chlorophenylmethylsulfone, aniline, 2-chloro-4-methylsulfonyl
VLMRGLCBIFWPGL-UHFFFAOYSA-N
2-chloro-4-methylsulfonylaniline
25801
95%
13244-35-4
100.0
95%
C7H8ClNO2S
2.5g
Solubility in water: negligible
CS(=O)(=O)C1=CC(=C(C=C1)N)Cl
205.66
205.66
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 236-225-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only