Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-(methylsulfonyl)benzoic acid, 96%

$68.86 - $68.86

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-(methylsulfonyl)benzoic acid
CAS 53250-83-2
Molecular Formula C8H7ClO4S
Formula Weight 234.66
MDL Number MFCD00216496
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2531903

  SDS
alfa aesar™
B2531903
1g Each for $68.86

AAB2531988

  SDS
alfa aesar™
B2531988
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Chloro-4-(methylsulfonyl)benzoic acid
Melting Point 191° to 196°C
Quantity 1g
CAS 53250-83-2
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280i-P305+P351+P338
Wear eye/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 96%
Molecular Formula C8H7ClO4S
Formula Weight 234.66
MDL Number MFCD00216496
Safety and Handling

Danger

GHS P Statement
P280i-P305+P351+P338
Wear eye/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H318
Causes serious eye damage.