missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4-methylpyrimidine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC453340010

 View more versions of this product

Catalog No. AC453340010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloro-4-methylpyrimidine
93°C to 95°C (17.0mmHg)
1g
97.5% min. (GC)
2-chloro-4-methyl pyrimidine, 2-chloro-4-methyl-pyrimidine, 2-chloro-6-methylpyrimidine, 4-methyl-2-chloro-pyrimidine, 4-methyl-2-chloropyrimidine, 4-methyl-2-pyrimidinylchloride, chloro-6 methylpyrimidine, ksc493s3l, pubchem6906, pyrimidine, 2-chloro-4-methyl
CC1=NC(=NC=C1)Cl
128.56
128.56
Glass Bottle
51°C to 56°C
13036-57-2
C5H5ClN2
BHAKRVSCGILCEW-UHFFFAOYSA-N
2-chloro-4-methylpyrimidine
11629607
98%

Chemical Identifiers

13036-57-2
128.56
2-chloro-4-methyl pyrimidine, 2-chloro-4-methyl-pyrimidine, 2-chloro-6-methylpyrimidine, 4-methyl-2-chloro-pyrimidine, 4-methyl-2-chloropyrimidine, 4-methyl-2-pyrimidinylchloride, chloro-6 methylpyrimidine, ksc493s3l, pubchem6906, pyrimidine, 2-chloro-4-methyl
2-chloro-4-methylpyrimidine
C5H5ClN2
BHAKRVSCGILCEW-UHFFFAOYSA-N
11629607
CC1=NC(=NC=C1)Cl
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents