missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-(methylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H55778MD

 View more versions of this product

Catalog No. AAH55778MD



Specifications

Specifications

2-Chloro-4-(methylcarbamoyl)benzeneboronic acid
1451391-89-1
MFCD22125174
LSMHGCJWZPHACP-UHFFFAOYSA-N
[2-chloro-4-(methylcarbamoyl)phenyl]boronic acid
73996177
97%
250mg
C8H9BClNO3
2-chloro-4-methylcarbamoyl benzeneboronic acid
B(C1=C(C=C(C=C1)C(=O)NC)Cl)(O)O
213.424
213.43

Chemical Identifiers

1451391-89-1
213.424
LSMHGCJWZPHACP-UHFFFAOYSA-N
73996177
B(C1=C(C=C(C=C1)C(=O)NC)Cl)(O)O
C8H9BClNO3
MFCD22125174
2-chloro-4-methylcarbamoyl benzeneboronic acid
[2-chloro-4-(methylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents