missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-(methylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5577803

 View more versions of this product

Catalog No. AAH5577803



Specifications

Specifications

2-Chloro-4-(methylcarbamoyl)benzeneboronic acid
1451391-89-1
MFCD22125174
LSMHGCJWZPHACP-UHFFFAOYSA-N
[2-chloro-4-(methylcarbamoyl)phenyl]boronic acid
73996177
97%
1g
C8H9BClNO3
2-chloro-4-methylcarbamoyl benzeneboronic acid
B(C1=C(C=C(C=C1)C(=O)NC)Cl)(O)O
213.424
213.43

Chemical Identifiers

1451391-89-1
213.424
LSMHGCJWZPHACP-UHFFFAOYSA-N
73996177
B(C1=C(C=C(C=C1)C(=O)NC)Cl)(O)O
C8H9BClNO3
MFCD22125174
2-chloro-4-methylcarbamoyl benzeneboronic acid
[2-chloro-4-(methylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents