Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-methylbenzotrifluoride, 97+%

$294.00 - $883.54

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-methylbenzotrifluoride
CAS 74483-46-8
Molecular Formula C8H6ClF3
Formula Weight 194.58
MDL Number MFCD00190112
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2348006

 
alfa aesar™
B2348006
5g Each for $294.00

AAB2348014

 
alfa aesar™
B2348014
25g Each for $883.54
Description & Specifications

Specifications

Formula Weight 194.58
Chemical Name or Material 2-Chloro-4-methylbenzotrifluoride
Assay ≥97%
Molecular Formula C8H6ClF3
MDL Number MFCD00190112
CAS 74483-46-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.