Please login to your online account to display your discounted pricing

2-chloro-4-methylaniline, 98%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC154671000

 View more versions of this product

Catalog No. AC154671000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 141.6
Quantity 100mL
Color Clear yellow to pale brown
Density 1.1510g/mL
Beilstein 12, 989
CAS Max % 100.0
Assay Percent Range 98%
CAS 615-65-6
Chemical Name or Material 2-Chloro-4-methylaniline
Molecular Formula C7H8ClN
Linear Formula ClC6H3(CH3)NH2
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
MDL Number MFCD00007666
Packaging Glass bottle
Refractive Index 1.5740 to 1.5760 (20°C, 589nm)
Storage Information Product may darken during storage
Infrared Spectrum Authentic
Synonym 2-Chloro-p-toluidine
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-Chloro-4-methylaniline
CAS 615-65-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8ClN
Linear Formula ClC6H3(CH3)NH2
Formula Weight 141.6
MDL Number MFCD00007666
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.

SDS & Safety Documents