Learn More
2-chloro-4-methylaniline, 98%, Thermo Scientific Chemicals
$215.69 - $215.69
Chemical Identifiers
CAS | 615-65-6 |
---|---|
Molecular Formula | C7H8ClN |
Molecular Weight (g/mol) | 141.6 |
MDL Number | MFCD00007666 |
InChI Key | XGYLSRFSXKAYCR-UHFFFAOYSA-N |
Synonym | 2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene |
PubChem CID | 12007 |
IUPAC Name | 2-chloro-4-methylaniline |
SMILES | CC1=CC(=C(C=C1)N)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC154670250
|
Thermo Scientific Chemicals
154670250 |
25 mL | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
615-65-6 | |
141.6 | |
XGYLSRFSXKAYCR-UHFFFAOYSA-N | |
12007 | |
CC1=CC(=C(C=C1)N)Cl |
C7H8ClN | |
MFCD00007666 | |
2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene | |
2-chloro-4-methylaniline |
Specifications
615-65-6 | |
100.0 | |
Brown to Yellow | |
223°C to 225°C | |
Authentic | |
Glass bottle | |
1.5740 to 1.576 | |
MFCD00007666 | |
12, 989 | |
2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene | |
CC1=CC(=C(C=C1)N)Cl | |
141.6 | |
141.6 | |
Liquid |
97.5 | |
7°C | |
1.1510g/mL | |
99°C | |
97.5% min. (GC) | |
C7H8ClN | |
ClC6H3(CH3)NH2 | |
25 mL | |
1.151 | |
XGYLSRFSXKAYCR-UHFFFAOYSA-N | |
2-chloro-4-methylaniline | |
12007 | |
98% | |
2-Chloro-4-methylaniline |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin t
GHS Signal Word: Danger
EINECSNumber : 210-440-7
Recommended Storage : Product may darken during storage
RUO – Research Use Only