missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-4-methylaniline, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC154670050

 View more versions of this product

Catalog No. AC154670050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloro-4-methylaniline
Authentic
1.5740 to 1.5760 (20°C, 589nm)
5mL
97.5
98%
ClC6H3(CH3)NH2
12, 989
XGYLSRFSXKAYCR-UHFFFAOYSA-N
2-chloro-4-methylaniline
12007
Liquid
1.1510g/mL
Glass bottle
Brown to Yellow
615-65-6
100.0
C7H8ClN
MFCD00007666
2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene
CC1=CC(=C(C=C1)N)Cl
141.598
141.6
≥97.5%

Chemical Identifiers

615-65-6
141.598
XGYLSRFSXKAYCR-UHFFFAOYSA-N
12007
CC1=CC(=C(C=C1)N)Cl
C7H8ClN
MFCD00007666
2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene
2-chloro-4-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : Product may darken during storage

SDS Documents