Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-methyl-3-nitropyridine, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-methyl-3-nitropyridine
CAS 23056-39-5
Molecular Formula C6H5ClN2O2
Formula Weight 172.57
MDL Number MFCD00012347
UN Number UN2811
Synonym 2-Chloro-3-nitro-4-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2605703

  SDS
alfa aesar™
H2605703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2605706

  SDS
alfa aesar™
H2605706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.57
Synonym 2-Chloro-3-nitro-4-picoline
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Chloro-4-methyl-3-nitropyridine
MDL Number MFCD00012347
CAS 23056-39-5
Melting Point 52° to 54°C
UN Number UN2811
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point >110°C (230°F)
Assay 97%
Molecular Formula C6H5ClN2O2
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.