Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-methoxyphenylacetonitrile, 95%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-methoxyphenylacetonitrile
CAS 170737-93-6
Molecular Formula C9H8ClNO
Formula Weight 181.62
MDL Number MFCD08445710
UN Number UN3439
Synonym 2-Chloro-4-methoxybenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2602103

  SDS
alfa aesar™
H2602103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2602106

  SDS
alfa aesar™
H2602106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.62
MDL Number MFCD08445710
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 2-Chloro-4-methoxyphenylacetonitrile
Health Hazard 1 Warning
Melting Point 41° to 43°C
CAS 170737-93-6
Molecular Formula C9H8ClNO
UN Number UN3439
Quantity 1g
Assay 95%
Health Hazard 3 GHS P Statement
P261-P280h-P305+P351+P338-P304+P340-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 2-Chloro-4-methoxybenzyl cyanide
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Applications
2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and acids.

Safety and Handling

Warning

GHS P Statement
P261-P280h-P305+P351+P338-P304+P340-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.