Learn More
2-Chloro-4-methoxyphenylacetonitrile, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2602103
Description
2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents and acids.
Specifications
2-Chloro-4-methoxyphenylacetonitrile | |
41°C to 43°C | |
MFCD08445710 | |
UN3439 | |
Insoluble in water. | |
COC1=CC(Cl)=C(CC#N)C=C1 | |
181.62 | |
181.62 |
170737-93-6 | |
C9H8ClNO | |
1 g | |
2-2-chloro-4-methoxyphenyl acetonitrile, 2-chloro-4-methoxyphenyl acetonitrile, 2-chloro-4-methoxyphenylacetonitrile, acmc-20appn, pubchem11235, 2-chloro-4-methoxyphenylacetnitrile, benzeneacetonitrile, 2-chloro-4-methoxy, 2-2-chloranyl-4-methoxy-phenyl ethanenitrile | |
UKYXNWLFQOJYNQ-UHFFFAOYSA-N | |
2-(2-chloro-4-methoxyphenyl)acetonitrile | |
22327506 | |
95% |
Chemical Identifiers
170737-93-6 | |
181.62 | |
UKYXNWLFQOJYNQ-UHFFFAOYSA-N | |
22327506 | |
COC1=CC(Cl)=C(CC#N)C=C1 |
C9H8ClNO | |
MFCD08445710 | |
2-2-chloro-4-methoxyphenyl acetonitrile, 2-chloro-4-methoxyphenyl acetonitrile, 2-chloro-4-methoxyphenylacetonitrile, acmc-20appn, pubchem11235, 2-chloro-4-methoxyphenylacetnitrile, benzeneacetonitrile, 2-chloro-4-methoxy, 2-2-chloranyl-4-methoxy-phenyl ethanenitrile | |
2-(2-chloro-4-methoxyphenyl)acetonitrile |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only