Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-iodopyridine, 99%

$72.04 - $238.59

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-iodopyridine
CAS 153034-86-7
Molecular Formula C5H3ClIN
Formula Weight 239.44
MDL Number MFCD01861983
Beilstein 108668
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAL1962303

 
alfa aesar™
L1962303
Each for $72.04

AAL1962306

 
alfa aesar™
L1962306
Each for $238.59
Description & Specifications

Specifications

Formula Weight 239.44
Chemical Name or Material 2-Chloro-4-iodopyridine
Assay 99%
Molecular Formula C5H3ClIN
CAS 153034-86-7
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD01861983
Beilstein 108668
Melting Point 42° to 44°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.