Learn More
2-Chloro-4-iodopyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1962303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloro-4-iodopyridine | |
42°C to 44°C | |
MFCD01861983 | |
108668 | |
2-chloro-4-iodo-pyridine, 4-iodo-2-chloropyridine, 2-chloro-iodopyridine, pyridine, 2-chloro-4-iodo, zlchem 826, pubchem2579, acmc-209d8n, 2-chloro-4-iodo pyridine, ksc174s0f, 2-chloro-4-iodopyridine | |
C1=CN=C(C=C1I)Cl | |
239.44 | |
239.44 |
153034-86-7 | |
C5H3ClIN | |
1 g | |
Light sensitive | |
KJKIPRQNFDUULB-UHFFFAOYSA-N | |
2-chloro-4-iodopyridine | |
1516511 | |
99% |
Chemical Identifiers
153034-86-7 | |
239.44 | |
KJKIPRQNFDUULB-UHFFFAOYSA-N | |
1516511 | |
C1=CN=C(C=C1I)Cl |
C5H3ClIN | |
MFCD01861983 | |
2-chloro-4-iodo-pyridine, 4-iodo-2-chloropyridine, 2-chloro-iodopyridine, pyridine, 2-chloro-4-iodo, zlchem 826, pubchem2579, acmc-209d8n, 2-chloro-4-iodo pyridine, ksc174s0f, 2-chloro-4-iodopyridine | |
2-chloro-4-iodopyridine |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
H302+H332-H315-H317-H318-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only