missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-iodonicotinic acid, 98%

$236.35 - $1,045.65

Chemical Identifiers

CAS 544671-78-5
Molecular Formula C6H3ClINO2
Formula Weight 283.45
MDL Number MFCD05663520
Chemical Name or Material 2-Chloro-4-iodonicotinic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5004903 SDS Alfa Aesar™
H5004903
1g Each for $236.35
AAH5004906 SDS Alfa Aesar™
H5004906
5g Each for $1,045.65
Specifications

Description

Applications
2-Chloro-4-iodonicotinic acid is used as a pharmaceutical intermediates.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

544671-78-5
283.45
2-Chloro-4-iodonicotinic acid
C6H3ClINO2
MFCD05663520

Specifications

162° to 167°C
544671-78-5
283.45
2-Chloro-4-iodonicotinic acid
1g
C6H3ClINO2
MFCD05663520
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning