Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine, tech. 90%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine
CAS 505084-55-9
Purity Grade Technical Grade
Formula Weight 307.44
MDL Number MFCD01862099
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6334503

  SDS
alfa aesar™
H6334503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6334506

  SDS
alfa aesar™
H6334506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63345MD

  SDS
alfa aesar™
H63345MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 307.44
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Purity Grade Technical Grade
Chemical Name or Material 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine
Physical Form Solid
MDL Number MFCD01862099
CAS 505084-55-9
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Sensitivity Light Sensitive
Assay 90%
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.