missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine, tech. 90%

Manufacturer:  Alfa Aesar™ H6334506

 View more versions of this product

Catalog No. AAH6334506



Specifications

Specifications

2-Chloro-4-iodo-5-(trifluoromethyl)pyridine
5g
Technical
MFCD01862099
XEEQBBXEZSNIQV-UHFFFAOYSA-N
2-chloro-4-iodo-5-(trifluoromethyl)pyridine
2773791
Solid
Light Sensitive
505084-55-9
C6H2ClF3IN
pyridine 2-chloro-4-iodo-5-trifluoromethyl, pyridine2-chloro-4-iodo-5-trifluoromethyl, 2-chloro-4-iodo-5-trifluoromethyl pyridine, 2-chloro-5-trifluoromethyl-4-iodopyridine, 2-chloro-4-iodo-5-trifluoromethyl-pyridine, acmc-1avx5, ksc496i6b
C1=C(C(=CN=C1Cl)C(F)(F)F)I
307.439
307.44
90%

Chemical Identifiers

505084-55-9
307.439
XEEQBBXEZSNIQV-UHFFFAOYSA-N
2773791
C1=C(C(=CN=C1Cl)C(F)(F)F)I
C6H2ClF3IN
MFCD01862099
pyridine 2-chloro-4-iodo-5-trifluoromethyl, pyridine2-chloro-4-iodo-5-trifluoromethyl, 2-chloro-4-iodo-5-trifluoromethyl pyridine, 2-chloro-5-trifluoromethyl-4-iodopyridine, 2-chloro-4-iodo-5-trifluoromethyl-pyridine, acmc-1avx5, ksc496i6b
2-chloro-4-iodo-5-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents