missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-hydroxybenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ H5577106

 View more versions of this product

Catalog No. AAH5577106



Description

Description

Applications
2-Chloro-4-hydroxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in organic solvents.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

2-Chloro-4-hydroxybenzaldehyde
145°C to 147°C
56962-11-9
MFCD00052184
Soluble in organic solvents.
C1=CC(=C(C=C1O)Cl)C=O
156.565
156.57
Air Sensitive
5g
C7H5ClO2
2-chlor-4-hydroxy-benzaldehyd
ZMOMCILMBYEGLD-UHFFFAOYSA-N
2-chloro-4-hydroxybenzaldehyde
185363
97%

Chemical Identifiers

56962-11-9
156.565
ZMOMCILMBYEGLD-UHFFFAOYSA-N
185363
C1=CC(=C(C=C1O)Cl)C=O
C7H5ClO2
MFCD00052184
2-chlor-4-hydroxy-benzaldehyd
2-chloro-4-hydroxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents