Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-hydroxy-6-methyl-5-nitropyrimidine, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-hydroxy-6-methyl-5-nitropyrimidine
CAS 65224-66-0
Formula Weight 189.56
MDL Number MFCD03844697
Synonym 2-Chloro-6-methyl-5-nitropyrimidin-4-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6395503

 
alfa aesar™
H6395503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6395506

 
alfa aesar™
H6395506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63955MD

 
alfa aesar™
H63955MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.56
Chemical Name or Material 2-Chloro-4-hydroxy-6-methyl-5-nitropyrimidine
Physical Form Solid
Assay 97%
MDL Number MFCD03844697
CAS 65224-66-0
Synonym 2-Chloro-6-methyl-5-nitropyrimidin-4-ol
Storage Note 1 2° to 8°C
Color Gray
Quantity 1g
Safety and Handling

2° to 8°C