missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-hydroxy-6-methyl-5-nitropyrimidine, 97%

Manufacturer:  Alfa Aesar™ H6395506

 View more versions of this product

Catalog No. AAH6395506



Specifications

Specifications

2-Chloro-4-hydroxy-6-methyl-5-nitropyrimidine
5g
C5H4ClN3O3
2-chloro-6-methyl-5-nitro-4 1h-pyrimidinone, 2-chloro-6-methyl-5-nitropyrimidin-4-ol, 2-chloro-6-methylnitro-4 1h-pyrimidinone, 2-chloro-6-methyl-5-nitrouracil, 2-chloro-6-methyl-5-nitro-3h-pyrimidin-4-one, 4 3h-pyrimidinone,2-chloro-6-methyl-5-nitro
CC1=C(C(=O)N=C(N1)Cl)[N+](=O)[O-]
189.555
189.56
97%
Gray
65224-66-0
MFCD03844697
GNXOLOBSWUZOEF-UHFFFAOYSA-N
2-chloro-6-methyl-5-nitro-1H-pyrimidin-4-one
12012221
Solid

Chemical Identifiers

65224-66-0
189.555
GNXOLOBSWUZOEF-UHFFFAOYSA-N
12012221
CC1=C(C(=O)N=C(N1)Cl)[N+](=O)[O-]
C5H4ClN3O3
MFCD03844697
2-chloro-6-methyl-5-nitro-4 1h-pyrimidinone, 2-chloro-6-methyl-5-nitropyrimidin-4-ol, 2-chloro-6-methylnitro-4 1h-pyrimidinone, 2-chloro-6-methyl-5-nitrouracil, 2-chloro-6-methyl-5-nitro-3h-pyrimidin-4-one, 4 3h-pyrimidinone,2-chloro-6-methyl-5-nitro
2-chloro-6-methyl-5-nitro-1H-pyrimidin-4-one
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents