missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4-fluoropyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 34941-91-8
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD04039345
InChI Key FGSAQRJRWCZLOB-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-4-fluoro, 2-chloro-4-fluoro pyridine, 2-chloro-4-fluoro-pyridine, pubchem6180, acmc-1aft7, 2-chlro-4-fluoroxypyridine, ksc222e0n, pyrimidine, 4-chloro-6-fluoro, abbypharma ap-14-5731
PubChem CID 2782792
IUPAC Name 2-chloro-4-fluoropyridine
SMILES C1=CN=C(C=C1F)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC442150050 SDS View Documents Acros Organics
AC442150050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC442150010 SDS View Documents Acros Organics
AC442150010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

34941-91-8
131.534
FGSAQRJRWCZLOB-UHFFFAOYSA-N
2782792
C1=CN=C(C=C1F)Cl
C5H3ClFN
MFCD04039345
pyridine, 2-chloro-4-fluoro, 2-chloro-4-fluoro pyridine, 2-chloro-4-fluoro-pyridine, pubchem6180, acmc-1aft7, 2-chlro-4-fluoroxypyridine, ksc222e0n, pyrimidine, 4-chloro-6-fluoro, abbypharma ap-14-5731
2-chloro-4-fluoropyridine

Specifications

1.456g/cm3
Colorless to Yellow
34941-91-8
MFCD04039345
FGSAQRJRWCZLOB-UHFFFAOYSA-N
2-chloro-4-fluoropyridine
2782792
Liquid
Glass bottle
5g
C5H3ClFN
pyridine, 2-chloro-4-fluoro, 2-chloro-4-fluoro pyridine, 2-chloro-4-fluoro-pyridine, pubchem6180, acmc-1aft7, 2-chlro-4-fluoroxypyridine, ksc222e0n, pyrimidine, 4-chloro-6-fluoro, abbypharma ap-14-5731
C1=CN=C(C=C1F)Cl
131.534
131.54