Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-fluorophenol, 98+%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-fluorophenol
CAS 1996-41-4
Molecular Formula C6H4ClFO
Formula Weight 146.55
MDL Number MFCD00002168
UN Number UN3265
Beilstein 1861277
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1086406

  SDS
alfa aesar™
A1086406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1086414

  SDS
alfa aesar™
A1086414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1086422

  SDS
alfa aesar™
A1086422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 146.55
Beilstein 1861277
Chemical Name or Material 2-Chloro-4-fluorophenol
Assay ≥98%
Molecular Formula C6H4ClFO
MDL Number MFCD00002168
CAS 1996-41-4
UN Number UN3265
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280-P273-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point 75°C (167°F)
Odor Phenol-like
Boiling Point 171° to 172°C
Melting Point 20° to 23°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P273-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.