Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-fluorocinnamic acid, 97%

$510.80 - $510.80

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-fluorocinnamic acid
CAS 133220-86-7
Molecular Formula C9H6ClFO2
Formula Weight 200.6
MDL Number MFCD00236027
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2306214

  SDS
alfa aesar™
B2306214
25g Each for $510.80
Description & Specifications

Specifications

Formula Weight 200.6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
MDL Number MFCD00236027
Chemical Name or Material 2-Chloro-4-fluorocinnamic acid
Quantity 25g
CAS 133220-86-7
Molecular Formula C9H6ClFO2
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 245° to 249°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.