Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-fluorobenzyl chloride, 96%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-fluorobenzyl chloride
CAS 93286-22-7
Molecular Formula C7H5Cl2F
Formula Weight 179.02
MDL Number MFCD00236026
UN Number UN3265
Beilstein 9043128
Synonym alpha,2-Dichloro-4-fluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1385006

  SDS
alfa aesar™
L1385006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1385014

  SDS
alfa aesar™
L1385014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1385022

  SDS
alfa aesar™
L1385022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.02
MDL Number MFCD00236026
Chemical Name or Material 2-Chloro-4-fluorobenzyl chloride
Molecular Formula C7H5Cl2F
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Synonym alpha,2-Dichloro-4-fluorotoluene
Health Hazard 1 Danger
CAS 93286-22-7
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 5g
UN Number UN3265
Beilstein 9043128
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay 96%

Applications
2-Chloro-4-fluorobenzyl chloride is used as a pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.