missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-fluorobenzyl chloride, 96%

Manufacturer:  Alfa Aesar™ L1385022

 View more versions of this product

Catalog No. AAL1385022



Description

Description

Applications
2-Chloro-4-fluorobenzyl chloride is used as a pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

2-Chloro-4-fluorobenzyl chloride
93286-22-7
MFCD00236026
9043128
HMIAXVWVTBIGON-UHFFFAOYSA-N
2-chloro-1-(chloromethyl)-4-fluorobenzene
2725064
96%
100g
C7H5Cl2F
UN3265
2-chloro-4-fluorobenzyl chloride, 2-chloro-1-chloromethyl-4-fluorobenzene, 2-chloro-4-fluorochlorobenzyl, 2-chloro-1-chloromethyl-4-fluoro-benzene, 2-chloro-4-fluorobenzylchloride, benzene, 2-chloro-1-chloromethyl-4-fluoro, alpha,2-dichloro-4-fluorotoluene, acmc-209rkz, ksc493q6n, paragos 500065
C1=CC(=C(C=C1F)Cl)CCl
179.015
179.02

Chemical Identifiers

93286-22-7
179.015
HMIAXVWVTBIGON-UHFFFAOYSA-N
2725064
C1=CC(=C(C=C1F)Cl)CCl
C7H5Cl2F
MFCD00236026
2-chloro-4-fluorobenzyl chloride, 2-chloro-1-chloromethyl-4-fluorobenzene, 2-chloro-4-fluorochlorobenzyl, 2-chloro-1-chloromethyl-4-fluoro-benzene, 2-chloro-4-fluorobenzylchloride, benzene, 2-chloro-1-chloromethyl-4-fluoro, alpha,2-dichloro-4-fluorotoluene, acmc-209rkz, ksc493q6n, paragos 500065
2-chloro-1-(chloromethyl)-4-fluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents