missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-4-fluorobenzyl bromide, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC298111000

 View more versions of this product

Catalog No. AC298111000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Chloro-4-fluorobenzyl bromide
White to yellow
45767-66-6
100.0
≥98.5%
C7H5BrClF
223.47
Authentic
100g
98.5
Crystalline Low Melting Mass
99%
F(Cl)C6H3CH2Br
MFCD00236025

Chemical Identifiers

45767-66-6
100.0
C7H5BrClF
223.47
2-Chloro-4-fluorobenzyl bromide
98.5
99%
F(Cl)C6H3CH2Br
MFCD00236025
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger