Please login to your online account to display your discounted pricing

2-chloro-4-fluorobenzyl bromide, 99%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  298110250

 View more versions of this product

Catalog No. AC298110250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Low Melting Mass
Formula Weight 223.47
Quantity 25g
Color White to yellow
CAS Max % 100.0
Chemical Name or Material 2-Chloro-4-fluorobenzyl bromide
Assay Percent Range 99%
CAS 45767-66-6
Molecular Formula C7H5BrClF
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
MDL Number MFCD00236025
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Linear Formula F(Cl)C6H3CH2Br

Specifications

Chemical Name or Material 2-Chloro-4-fluorobenzyl bromide
CAS 45767-66-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H5BrClF
Linear Formula F(Cl)C6H3CH2Br
Formula Weight 223.47
MDL Number MFCD00236025
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

SDS & Safety Documents