Please login to your online account to display your discounted pricing

2-chloro-4-fluorobenzoyl Chloride, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC382000010

 View more versions of this product

Catalog No. AC382000010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 193
Quantity 1g
Color Clear pale yellow to yellow
CAS Max % 100.0
Linear Formula FC6H3(Cl)COCl
Health Hazard 1 GHS Signal Word: Danger
CAS 21900-54-9
MDL Number MFCD00084954
Assay Percent Range 98%
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C7 H3 Cl2 F O
CAS Min % 97.5
Infrared Spectrum Authentic
Refractive Index 1.549 to 1.551 (20°C, 589nm)
Chemical Name or Material 2-Chloro-4-fluorobenzoyl chloride
Synonym 2-Chloro-4-fluorobenzene-1-carbonyl chloride
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

Specifications

Chemical Name or Material 2-Chloro-4-fluorobenzoyl chloride
CAS 21900-54-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H3 Cl2 F O
Linear Formula FC6H3(Cl)COCl
Formula Weight 193
MDL Number MFCD00084954
Synonym 2-Chloro-4-fluorobenzene-1-carbonyl chloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents