missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-4-fluorobenzoyl Chloride, 98%, ACROS Organics™

Chemical Identifiers

CAS 21900-54-9
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7 H3 Cl2 F O
Linear Formula FC6H3(Cl)COCl
Formula Weight 193
MDL Number MFCD00084954
Synonym 2-Chloro-4-fluorobenzene-1-carbonyl chloride
Chemical Name or Material 2-Chloro-4-fluorobenzoyl chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC382000010 SDS Thermo Fisher Scientific
AC382000010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

21900-54-9
100.0
C7 H3 Cl2 F O
193
2-Chloro-4-fluorobenzene-1-carbonyl chloride
≥97.5%
97.5
98%
FC6H3(Cl)COCl
MFCD00084954
2-Chloro-4-fluorobenzoyl chloride

Specifications

Authentic
1g
97.5
100.0
Liquid
C7 H3 Cl2 F O
193
2-Chloro-4-fluorobenzene-1-carbonyl chloride
≥97.5%
Clear pale yellow to yellow
21900-54-9
Glass bottle
1.549 to 1.551 (20°C, 589nm)
98%
FC6H3(Cl)COCl
MFCD00084954
2-Chloro-4-fluorobenzoyl chloride

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger