Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-fluorobenzenesulfonyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-fluorobenzenesulfonyl chloride
CAS 85958-57-2
Molecular Formula C6H3Cl2FO2S
Formula Weight 229.06
MDL Number MFCD00052957
UN Number UN3265
Beilstein 5520034
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1053603

  SDS
alfa aesar™
A1053603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.06
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Melting Point 28° to 31°C
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Beilstein 5520034
Molecular Formula C6H3Cl2FO2S
UN Number UN3265
Assay 98%
Flash Point >110°C (230°F)
Boiling Point 243° to 244°C
MDL Number MFCD00052957
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Chemical Name or Material 2-Chloro-4-fluorobenzenesulfonyl chloride
CAS 85958-57-2
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.