Please login to your online account to display your discounted pricing

2-chloro-4-fluorobenzene-1-sulfonyl chloride, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  TL00318DA

 View more versions of this product

Catalog No. TL00318DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 229.06
Quantity 1g
Assay 97%
CAS Min % 97.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Chemical Name or Material 2-Chloro-4-fluorobenzene-1-sulfonyl chloride
Molecular Formula C6H3Cl2FO2S
CAS 85958-57-2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Linear Formula Cl(F)C6H3(SO2Cl)
MDL Number MFCD00052957
Assay Percent Range 97%

Specifications

Chemical Name or Material 2-Chloro-4-fluorobenzene-1-sulfonyl chloride
CAS 85958-57-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H3Cl2FO2S
Linear Formula Cl(F)C6H3(SO2Cl)
Formula Weight 229.06
MDL Number MFCD00052957
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents