Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-fluorobenzaldehyde, 97%

$44.89 - $625.31

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-fluorobenzaldehyde
CAS 84194-36-5
Molecular Formula C7H4ClFO
Formula Weight 158.56
MDL Number MFCD00042527
Beilstein 3537704
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1534503

  SDS
alfa aesar™
A1534503
1g Each for $44.89

AAA1534506

  SDS
alfa aesar™
A1534506
5g Each for $156.70

AAA1534514

  SDS
alfa aesar™
A1534514
25g Each for $625.31
Description & Specifications

Specifications

Formula Weight 158.56
Quantity 1g
Molecular Formula C7H4ClFO
Beilstein 3537704
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 2-Chloro-4-fluorobenzaldehyde
Melting Point 60° to 63°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 118° to 120°C (50mmHg)
MDL Number MFCD00042527
Flash Point >110°C (230°F)
CAS 84194-36-5
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.