Please login to your online account to display your discounted pricing

2-Chloro-4'-fluoroacetophenone, 99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC158175000

 View more versions of this product

Catalog No. AC158175000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Flakes or Platelets
Assay ≥98.5%
Formula Weight 172.59
Quantity 500g
Color Light yellow to yellow-beige
Melting Point 47° to 50°C
CAS Max % 100.0
Synonym 2-Chloro-1-(4-fluorophenyl)ethanone
Chemical Name or Material 2-Chloro-4'-fluoroacetophenone
Molecular Formula C8H6ClFO
MDL Number MFCD00011652
CAS 456-04-2
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Wear respiratory protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula FC6H4COCH2Cl

Specifications

Chemical Name or Material 2-Chloro-4'-fluoroacetophenone
CAS 456-04-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H6ClFO
Linear Formula FC6H4COCH2Cl
Formula Weight 172.59
MDL Number MFCD00011652
Synonym 2-Chloro-1-(4-fluorophenyl)ethanone
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Wear respiratory protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents